The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022724.4 | NP_073561.2 | 2 - 3 | 425 | 75 |
GenBank mRNA | AF149205.1 | - | 2 - 3 | 403 | 75 |
AK083457.1 | - | 2 - 3 | 403 | 75 | |
CX227854.1 | - | 2 - 3 | 425 | 75 |