Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
67426
Gene Name
aarF domain containing kinase 3
Gene Aliases
4632432J16Rik, AI462003, Cabc1, mKIAA0451
Chromosome Location
Chr.1: 180165238 - 180196020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163290.1 NP_001156762.1 13 - 14 1624 122
NM_023341.3 NP_075830.2 13 - 14 1722 122
GenBank mRNA AJ278735.1 - 13 - 14 1602 122
AK004177.1 - 6 - 7 607 122
AK004791.1 - 13 - 14 1664 122
AK014605.2 - 13 - 14 1664 122
BC030937.1 - 13 - 14 1670 122

View Full Product Details