The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023712.3 | NP_076201.2 | 3 - 4 | 1302 | 103 |
GenBank mRNA | AF212372.1 | - | 3 - 4 | 793 | 103 |
AK154353.1 | - | 3 - 4 | 792 | 103 | |
AK166931.1 | - | 3 - 4 | 794 | 103 | |
BC002297.1 | - | 3 - 4 | 321 | 103 | |
BC026854.1 | - | 3 - 4 | 763 | 103 | |
BC085491.1 | - | 3 - 4 | 746 | 103 |