Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smtnl1
Entrez Gene ID
68678
Gene Name
smoothelin-like 1
Gene Aliases
1110030K22Rik, CHASM
Chromosome Location
Chr.2: 84811176 - 84826934 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024230.2 NP_077192.1 3 - 4 930 125
GenBank mRNA BC002317.1 - 3 - 4 930 125

View Full Product Details