Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf10
Entrez Gene ID
72057
Gene Name
PHD finger protein 10
Gene Aliases
1810055P05Rik, AV024533
Chromosome Location
Chr.17: 14944994 - 14961638 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024250.4 NP_077212.3 8 - 9 1021 91
GenBank mRNA AB221576.1 - 6 - 7 688 91
AK007873.1 - 8 - 9 1001 91
BC002206.1 - 6 - 7 755 91
CA322732.1 - 5 - 6 579 91
CD363469.1 - 2 - 3 278 91

View Full Product Details