The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026172.3 | NP_080448.1 | 2 - 3 | 428 | 59 |
GenBank mRNA | AK002756.1 | - | 2 - 3 | 308 | 59 |
AK004725.2 | - | 2 - 3 | 295 | 59 | |
AK015692.1 | - | 2 - 3 | 299 | 59 | |
AK146003.1 | - | 2 - 3 | 299 | 59 | |
AK168490.1 | - | 2 - 3 | 296 | 59 | |
BC046972.1 | - | 2 - 3 | 428 | 59 | |
CT010405.1 | - | 2 - 3 | 272 | 59 |