The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026302.3 | NP_080578.1 | 12 - 13 | 1179 | 60 |
GenBank mRNA | AK016059.1 | - | 12 - 13 | 1233 | 60 |
AK034292.1 | - | 13 - 14 | 1237 | 60 | |
AK141855.1 | - | 13 - 14 | 1203 | 60 | |
AK160813.1 | - | 12 - 13 | 1185 | 60 | |
AK163240.1 | - | 12 - 13 | 1201 | 60 | |
AK168484.1 | - | 12 - 13 | 1185 | 60 | |
BC006677.1 | - | 12 - 13 | 1142 | 60 | |
BC034725.1 | - | 12 - 13 | 1179 | 60 |