The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026417.4 | NP_080693.2 | 1 - 2 | 300 | 141 |
GenBank mRNA | AK009618.1 | - | 1 - 2 | 286 | 141 |
AK017721.1 | - | 1 - 2 | 278 | 141 | |
AK017722.1 | - | 1 - 2 | 278 | 141 | |
AK083301.1 | - | 1 - 2 | 280 | 141 | |
AK159227.1 | - | 1 - 2 | 261 | 141 | |
AK166347.1 | - | 1 - 2 | 288 | 141 | |
BY709789.1 | - | 1 - 2 | 300 | 141 |