Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgs10
Entrez Gene ID
67865
Gene Name
regulator of G-protein signalling 10
Gene Aliases
2310010N19Rik
Chromosome Location
Chr.7: 128373621 - 128419061 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026418.2 NP_080694.1 4 - 5 456 67
GenBank mRNA AK009283.1 - 4 - 5 457 67
AK009686.1 - 4 - 5 458 67
AK017314.1 - 4 - 5 457 67
AK088009.1 - 4 - 5 455 67
BC109178.1 - 4 - 5 441 67
BC109179.1 - 4 - 5 446 67

View Full Product Details