The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167829.1 | NP_001161301.1 | 5 - 6 | 640 | 107 |
NM_026437.3 | NP_080713.2 | 5 - 6 | 640 | 107 | |
GenBank mRNA | AK004643.1 | - | 4 - 5 | 560 | 107 |
AK007867.1 | - | 5 - 6 | 600 | 107 | |
AK145769.1 | - | 5 - 6 | 606 | 107 | |
AK168474.1 | - | 4 - 5 | 554 | 107 | |
BC013514.1 | - | 5 - 6 | 599 | 107 |