Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppa1
Entrez Gene ID
67895
Gene Name
pyrophosphatase (inorganic) 1
Gene Aliases
2010317E03Rik, Pyp
Chromosome Location
Chr.10: 61648621 - 61674165 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026438.4 NP_080714.2 10 - 11 942 114
GenBank mRNA AK008575.1 - 10 - 11 943 114
AK020291.1 - 10 - 11 958 114
AK088397.1 - 10 - 11 938 114
AK145640.1 - 10 - 11 992 114
AK146300.1 - 10 - 11 905 114
AK151512.1 - 10 - 11 933 114
BC010468.1 - 10 - 11 876 114

View Full Product Details