The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026551.3 | NP_080827.2 | 4 - 5 | 598 | 75 |
GenBank mRNA | AK002835.1 | - | 1 - 2 | 198 | 75 |
AK051022.1 | - | 4 - 5 | 599 | 75 | |
AK078460.1 | - | 4 - 5 | 531 | 75 | |
AK162069.1 | - | 4 - 5 | 558 | 75 | |
AK170762.1 | - | 4 - 5 | 549 | 75 | |
AK180504.1 | - | 1 - 2 | 79 | 75 | |
AK180764.1 | - | 1 - 2 | 79 | 75 | |
BC005607.1 | - | 4 - 5 | 564 | 75 | |
BC023208.1 | - | 3 - 4 | 512 | 75 | |
BC031855.1 | - | 4 - 5 | 547 | 75 | |
BC042685.1 | - | 3 - 4 | 512 | 75 | |
BC045193.1 | - | 4 - 5 | 552 | 75 | |
BC047340.1 | - | 3 - 4 | 512 | 75 |