The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026594.2 | NP_080870.1 | 2 - 3 | 175 | 84 |
GenBank mRNA | AK005645.1 | - | 2 - 3 | 165 | 84 |
AK019681.1 | - | 2 - 3 | 177 | 84 | |
BC023885.1 | - | 2 - 3 | 165 | 84 |