The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027397.3 | NP_081673.2 | 3 - 4 | 642 | 158 |
GenBank mRNA | AK013964.1 | - | 3 - 4 | 585 | 158 |