The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028889.2 | NP_083165.1 | 3 - 4 | 674 | 70 |
GenBank mRNA | AK016491.1 | - | 3 - 4 | 674 | 70 |
AK045911.1 | - | 3 - 4 | 676 | 70 | |
AK144175.1 | - | 3 - 4 | 677 | 70 | |
AK171846.1 | - | 3 - 4 | 676 | 70 | |
BC019531.1 | - | 3 - 4 | 660 | 70 | |
BC085088.1 | - | 3 - 4 | 659 | 70 |