The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029091.2 | NP_083367.1 | 6 - 7 | 956 | 98 |
GenBank mRNA | AK004774.1 | - | 6 - 7 | 957 | 98 |
AK170793.1 | - | 6 - 7 | 940 | 98 |