Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pelp1
- Entrez Gene ID
-
75273
- Gene Name
- proline, glutamic acid and leucine rich protein 1
- Gene Aliases
-
4930563C04Rik, MNAR
- Chromosome Location
-
Chr.11: 70392881 - 70410031 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016197.1 |
- |
3 - 4 |
436 |
86 |
View Full Product Details