The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289554.1 | NP_001276483.1 | 7 - 8 | 857 | 72 |
NM_001289555.1 | NP_001276484.1 | 7 - 8 | 854 | 72 | |
NM_001289556.1 | NP_001276485.1 | 6 - 7 | 698 | 72 | |
NM_001289557.1 | NP_001276486.1 | 5 - 6 | 549 | 72 | |
NM_029659.4 | NP_083935.3 | 7 - 8 | 864 | 72 | |
GenBank mRNA | AK005599.1 | - | 7 - 8 | 855 | 72 |
AK005858.1 | - | 6 - 7 | 605 | 72 | |
AK161310.1 | - | 7 - 8 | 865 | 72 | |
BC045148.1 | - | 7 - 8 | 794 | 72 | |
BF471066.1 | - | 5 - 6 | 458 | 72 |