The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030707.3 | NP_109632.2 | 6 - 7 | 1235 | 65 |
GenBank mRNA | AB028174.1 | - | 6 - 7 | 1266 | 65 |
AF302046.1 | - | 6 - 7 | 1304 | 65 | |
AF329486.1 | - | 6 - 7 | 1236 | 65 | |
AK034125.1 | - | 6 - 7 | 1236 | 65 | |
AK076123.1 | - | 6 - 7 | 1269 | 65 | |
AY506556.1 | - | 6 - 7 | 1275 | 65 | |
BC016551.1 | - | 6 - 7 | 1278 | 65 |