Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glis2
Entrez Gene ID
83396
Gene Name
GLIS family zinc finger 2
Gene Aliases
Gli5, Klf16, Nkl
Chromosome Location
Chr.16: 4594713 - 4625141 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031184.3 NP_112461.2 2 - 3 446 91
GenBank mRNA AF249340.1 - 1 - 2 228 91
AF325913.1 - 1 - 2 198 91
BC016124.1 - 1 - 2 195 91
BC021517.1 - 2 - 3 275 91
CF727873.1 - 2 - 3 446 91
CN703218.1 - 1 - 2 82 91
CX563691.1 - 1 - 2 123 91

View Full Product Details