Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Glis2
Entrez Gene ID
83396
Gene Name
GLIS family zinc finger 2
Gene Aliases
Gli5, Klf16, Nkl
Chromosome Location
Chr.16: 4594713 - 4625141 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031184.3 NP_112461.2 5 - 6 927 92
GenBank mRNA AF249340.1 - 4 - 5 706 89
AF325913.1 - 4 - 5 679 92
BC021517.1 - 5 - 6 756 92
CN703218.1 - 4 - 5 398 92
CX563691.1 - 4 - 5 604 92

View Full Product Details