The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031843.2 | NP_114031.2 | 8 - 9 | 966 | 60 |
GenBank mRNA | AF285235.1 | - | 8 - 9 | 961 | 60 |
AK206513.1 | - | 2 - 3 | 194 | 60 | |
BC027205.1 | - | 8 - 9 | 966 | 60 |