Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tas1r1
Entrez Gene ID
110326
Gene Name
taste receptor, type 1, member 1
Gene Aliases
Gpr70, T1r1, TR1
Chromosome Location
Chr.4: 152027914 - 152038490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031867.2 NP_114073.1 5 - 6 1701 73
GenBank mRNA AF301161.1 - 5 - 6 1597 73
AF301162.1 - 5 - 6 1597 73
AF337040.1 - 5 - 6 1779 73
AK153579.1 - 5 - 6 1651 73
AY032622.1 - 5 - 6 1597 73

View Full Product Details