The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032007.3 | NP_114396.3 | 5 - 6 | 804 | 117 |
GenBank mRNA | AJ278461.1 | - | 5 - 6 | 773 | 117 |