The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033573.2 | NP_291051.1 | 6 - 7 | 1611 | 83 |
GenBank mRNA | AF273750.1 | - | 6 - 7 | 1632 | 83 |
AK078394.1 | - | 6 - 7 | 1631 | 83 | |
BC052072.1 | - | 6 - 7 | 1611 | 83 | |
BC082784.1 | - | 6 - 7 | 1629 | 83 |