The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033603.2 | NP_291081.2 | 3 - 4 | 220 | 63 |
GenBank mRNA | AF320619.1 | - | 3 - 4 | 220 | 63 |
AK002318.1 | - | 3 - 4 | 219 | 63 | |
AK017204.1 | - | 3 - 4 | 234 | 63 | |
BC087954.1 | - | 3 - 4 | 233 | 63 |