Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fgd6
Entrez Gene ID
13998
Gene Name
FYVE, RhoGEF and PH domain containing 6
Gene Aliases
AA123052, Etohd4, ZFYVE24
Chromosome Location
Chr.10: 94036001 - 94145339 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053072.3 NP_444302.4 20 - 21 4460 90
GenBank mRNA AK016940.1 - 14 - 15 1433 90
AK173157.1 - 20 - 21 4300 90
BC026860.1 - 19 - 20 2798 90
BC139019.1 - 20 - 21 4254 90
BC139022.1 - 20 - 21 4254 90

View Full Product Details