The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053241.5 | NP_444471.1 | 4 - 5 | 927 | 66 |
GenBank mRNA | AB009016.1 | - | 4 - 5 | 589 | 66 |
AF323748.1 | - | 4 - 5 | 717 | 66 | |
AK085268.1 | - | 4 - 5 | 928 | 66 | |
AK085333.1 | - | 4 - 5 | 903 | 66 | |
BC115574.2 | - | 5 - 6 | 706 | 66 | |
BC115575.2 | - | 5 - 6 | 706 | 66 |