Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyve1
Entrez Gene ID
114332
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Gene Aliases
1200012G08Rik, Crsbp-1, Lyve-1, Xlkd1
Chromosome Location
Chr.7: 110850607 - 110862953 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053247.4 NP_444477.2 3 - 4 477 71
GenBank mRNA AJ311501.1 - 3 - 4 632 71
AK004726.2 - 3 - 4 477 71
BC038653.1 - 3 - 4 583 71
BC038892.1 - 3 - 4 583 71

View Full Product Details