The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007395.3 | NP_031421.1 | 8 - 9 | 1412 | 65 |
GenBank mRNA | AK154578.1 | - | - | 1435 | 65 |
AK157910.1 | - | 8 - 9 | 1413 | 65 | |
BC059832.1 | - | - | 1405 | 65 | |
BC145775.1 | - | 8 - 9 | 1465 | 65 | |
BC145777.1 | - | 8 - 9 | 1465 | 65 | |
Z31663.1 | - | 8 - 9 | 1401 | 65 |