Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap1m1
Entrez Gene ID
11767
Gene Name
adaptor-related protein complex AP-1, mu subunit 1
Gene Aliases
AA408894, AP47, Adtm1A, Cltnm, [m]1A, mu1A
Chromosome Location
Chr.8: 72240050 - 72257385 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007456.4 NP_031482.1 7 - 8 888 62
GenBank mRNA AK148173.1 - 7 - 8 903 62
BC003823.1 - 7 - 8 862 62
M62419.1 - 7 - 8 890 62

View Full Product Details