The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080999.2 | NP_001074468.1 | 6 - 7 | 1438 | 86 |
NM_001081000.2 | NP_001074469.1 | 6 - 7 | 1485 | 86 | |
NM_001195205.1 | NP_001182134.1 | 6 - 7 | 1438 | 86 | |
GenBank mRNA | AK080170.1 | - | 6 - 7 | 1313 | 86 |
AK133844.1 | - | 6 - 7 | 1313 | 86 | |
AK134368.1 | - | 6 - 7 | 1439 | 86 | |
AK146391.1 | - | 6 - 7 | 1378 | 86 | |
AK150482.1 | - | 6 - 7 | 1283 | 86 | |
AK151311.1 | - | 6 - 7 | 1283 | 86 | |
AK151422.1 | - | 6 - 7 | 1309 | 86 | |
AK151725.1 | - | 6 - 7 | 1283 | 86 | |
AK151778.1 | - | 6 - 7 | 1283 | 86 | |
AK152055.1 | - | 6 - 7 | 1283 | 86 | |
AK152201.1 | - | 6 - 7 | 1283 | 86 | |
AK152363.1 | - | 6 - 7 | 1283 | 86 | |
AK152548.1 | - | 6 - 7 | 1283 | 86 | |
AK152903.1 | - | 6 - 7 | 1283 | 86 | |
AK153043.1 | - | 6 - 7 | 1283 | 86 | |
AK153190.1 | - | 6 - 7 | 1309 | 86 | |
AK185507.1 | - | 3 - 4 | 340 | 86 | |
BC027431.1 | - | 5 - 6 | 1550 | 86 | |
BC038486.1 | - | 6 - 7 | 1311 | 86 | |
X56044.1 | - | 5 - 6 | 1882 | 86 |