The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290368.1 | NP_001277297.1 | 1 - 2 | 201 | 130 |
NM_001290369.1 | NP_001277298.1 | 1 - 2 | 201 | 130 | |
NM_001290370.1 | NP_001277299.1 | 1 - 2 | 201 | 130 | |
NM_008708.2 | NP_032734.1 | 1 - 2 | 201 | 130 | |
GenBank mRNA | AF043327.1 | - | 1 - 2 | 125 | 130 |
AK044213.1 | - | 1 - 2 | 133 | 130 | |
AK134521.1 | - | 1 - 2 | 165 | 130 | |
AK161490.1 | - | 1 - 2 | 201 | 130 | |
AK168171.1 | - | 1 - 2 | 169 | 130 | |
AK168190.1 | - | 1 - 2 | 169 | 130 | |
BC037647.1 | - | 1 - 2 | 148 | 130 |