The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008730.2 | NP_032756.1 | 4 - 5 | 1308 | 86 |
GenBank mRNA | AK140128.1 | - | 4 - 5 | 825 | 86 |
BC093479.1 | - | 4 - 5 | 1233 | 86 | |
BC139018.1 | - | 4 - 5 | 1215 | 86 | |
BC139020.1 | - | 4 - 5 | 1215 | 86 |