Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsg2
Entrez Gene ID
18197
Gene Name
neuron specific gene family member 2
Gene Aliases
-
Chromosome Location
Chr.11: 32000386 - 32059211 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290680.1 NP_001277609.1 2 - 3 193 77
NM_001290681.1 NP_001277610.1 2 - 3 189 77
NM_008741.4 NP_032767.1 2 - 3 329 77
GenBank mRNA AK011563.1 - 1 - 2 145 77
AK041338.1 - 2 - 3 247 77
AK049679.1 - 2 - 3 286 77
AK075612.1 - 2 - 3 286 77
AK088499.1 - 2 - 3 321 77
BC018224.1 - 2 - 3 261 77
BU058771.1 - 2 - 3 192 77
DN176718.1 - 2 - 3 329 77
U17259.1 - 2 - 3 289 77

View Full Product Details