Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nxn
Entrez Gene ID
18230
Gene Name
nucleoredoxin
Gene Aliases
l11Jus13
Chromosome Location
Chr.11: 76257226 - 76399141 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008750.5 NP_032776.1 3 - 4 699 90
GenBank mRNA AB095441.1 - 3 - 4 544 90
AB095442.1 - 3 - 4 544 90
AB095443.1 - 3 - 4 544 90
AB095444.1 - 3 - 4 544 90
AK012352.1 - 3 - 4 684 90
AK048109.1 - 3 - 4 698 90
AK149075.1 - 3 - 4 683 90
BC004688.1 - 3 - 4 659 90
BC047339.1 - 2 - 3 231 90
BC058244.1 - 3 - 4 699 90

View Full Product Details