Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcsk7
Entrez Gene ID
18554
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
AA959856, PC7, SPC7
Chromosome Location
Chr.9: 45906569 - 45929726 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281934.1 NP_001268863.1 2 - 3 860 67
NM_008794.2 NP_032820.2 2 - 3 596 67
NR_104055.1 2 - 3 596 67
GenBank mRNA AK148416.1 - 2 - 3 181 67
AK151980.1 - 2 - 3 183 67
AK165389.1 - 2 - 3 202 67
BC006730.1 - 1 - 2 97 67
BC007322.1 - 2 - 3 459 67
BC056456.1 - 2 - 3 596 67
U48830.1 - 2 - 3 168 67

View Full Product Details