The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281934.1 | NP_001268863.1 | 9 - 10 | 2023 | 73 |
NM_008794.2 | NP_032820.2 | 9 - 10 | 1759 | 73 | |
NR_104055.1 | 9 - 10 | 1759 | 73 | ||
GenBank mRNA | AK148416.1 | - | 9 - 10 | 1344 | 73 |
AK151980.1 | - | 9 - 10 | 1346 | 73 | |
AK165389.1 | - | 9 - 10 | 1365 | 73 | |
BC006730.1 | - | 8 - 9 | 1260 | 73 | |
BC007322.1 | - | 9 - 10 | 1622 | 73 | |
BC056456.1 | - | 9 - 10 | 1758 | 73 | |
U48830.1 | - | 9 - 10 | 1331 | 73 |