The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008942.2 | NP_032968.2 | 14 - 15 | 1721 | 120 |
GenBank mRNA | AK133898.1 | - | 14 - 15 | 1529 | 120 |
BC009653.1 | - | 14 - 15 | 1660 | 120 | |
BC086798.1 | - | 14 - 15 | 1713 | 120 | |
BC098212.1 | - | 14 - 15 | 1721 | 120 | |
U35646.1 | - | 14 - 15 | 1708 | 120 |