Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npepps
Entrez Gene ID
19155
Gene Name
aminopeptidase puromycin sensitive
Gene Aliases
AAP-S, MP100, Psa, R74825, goku
Chromosome Location
Chr.11: 97205842 - 97280638 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008942.2 NP_032968.2 22 - 23 2721 115
GenBank mRNA AK133898.1 - 22 - 23 2529 115
BC009653.1 - 22 - 23 2660 115
BC086798.1 - 22 - 23 2713 115
BC098212.1 - 22 - 23 2721 115
U35646.1 - 22 - 23 2708 115

View Full Product Details