The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008978.2 | NP_033004.1 | 3 - 4 | 251 | 102 |
GenBank mRNA | AK017003.1 | - | 3 - 4 | 314 | 102 |
AK029493.1 | - | 3 - 4 | 249 | 102 | |
BC139817.1 | - | 1 - 2 | 142 | 102 | |
D64141.1 | - | 2 - 3 | 250 | 102 |