The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271549.1 | NP_001258478.1 | 13 - 14 | 1660 | 75 |
NM_001271550.1 | NP_001258479.1 | 13 - 14 | 1651 | 75 | |
NM_001271551.1 | NP_001258480.1 | 13 - 14 | 1648 | 75 | |
NM_001271552.1 | NP_001258481.1 | 12 - 13 | 1486 | 75 | |
NM_001271553.1 | NP_001258482.1 | 12 - 13 | 1483 | 75 | |
NM_001271554.1 | NP_001258483.1 | 12 - 13 | 1474 | 75 | |
NM_001271555.1 | NP_001258484.1 | 12 - 13 | 1471 | 75 | |
NM_001271556.1 | NP_001258485.1 | 11 - 12 | 1364 | 75 | |
NM_001271557.1 | NP_001258486.1 | 11 - 12 | 1361 | 75 | |
NM_001271558.1 | NP_001258487.1 | 11 - 12 | 1352 | 75 | |
NM_001271559.1 | NP_001258488.1 | 11 - 12 | 1349 | 75 | |
NM_009646.2 | NP_033776.1 | 13 - 14 | 1663 | 75 | |
NR_073358.1 | 14 - 15 | 1718 | 75 | ||
GenBank mRNA | AF079536.1 | - | 13 - 14 | 1633 | 75 |
AF128115.1 | - | 13 - 14 | 1630 | 75 | |
AF128116.1 | - | 13 - 14 | 1621 | 75 | |
AF128117.1 | - | 13 - 14 | 1618 | 75 | |
AF128118.1 | - | 12 - 13 | 1456 | 75 | |
AF128119.1 | - | 12 - 13 | 1453 | 75 | |
AF128120.1 | - | 12 - 13 | 1444 | 75 | |
AF128121.1 | - | 12 - 13 | 1441 | 75 | |
AF128122.1 | - | 11 - 12 | 1334 | 75 | |
AF128123.1 | - | 11 - 12 | 1331 | 75 | |
AF128124.1 | - | 11 - 12 | 1322 | 75 | |
AF128125.1 | - | 11 - 12 | 1319 | 75 | |
AF128772.1 | - | 13 - 14 | 1576 | 75 | |
AF128773.1 | - | 13 - 14 | 1576 | 75 | |
AJ132243.1 | - | 13 - 14 | 1576 | 75 | |
AJ243821.1 | - | 13 - 14 | 1576 | 75 | |
AK131938.1 | - | 4 - 5 | 663 | 75 | |
AK153690.1 | - | 13 - 14 | 2663 | 75 | |
BC103510.1 | - | 14 - 15 | 1680 | 75 | |
BC103511.1 | - | 12 - 13 | 1448 | 75 | |
BC103518.1 | - | 13 - 14 | 1622 | 75 | |
EU625343.3 | - | 13 - 14 | 1593 | 75 | |
JF267399.1 | - | 13 - 14 | 1593 | 75 |