Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nfyb
Entrez Gene ID
18045
Gene Name
nuclear transcription factor-Y beta
Gene Aliases
AA985999, Cbf-A
Chromosome Location
Chr.10: 82748700 - 82764148 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010914.2 NP_035044.1 7 - 8 801 89
GenBank mRNA AK010762.1 - 7 - 8 781 89
AK146168.1 - 7 - 8 803 89
BC010719.1 - 7 - 8 742 89
X55316.1 - 7 - 8 797 89

View Full Product Details