The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025568.2 | NP_001020739.1 | 4 - 5 | 973 | 62 |
NM_001159952.1 | NP_001153424.1 | 3 - 4 | 946 | 62 | |
NM_001159953.1 | NP_001153425.1 | 4 - 5 | 973 | 62 | |
NM_001159955.1 | NP_001153427.1 | 5 - 6 | 535 | 62 | |
NM_001159956.1 | NP_001153428.1 | 3 - 4 | 392 | 62 | |
NM_001159957.1 | NP_001153429.1 | 4 - 5 | 419 | 62 | |
NM_001159960.1 | NP_001153432.1 | 5 - 6 | 847 | 62 | |
NM_011054.4 | NP_035184.1 | 5 - 6 | 950 | 62 | |
GenBank mRNA | AK014887.1 | - | 3 - 4 | 363 | 62 |
AK015375.1 | - | 5 - 6 | 504 | 62 | |
AK017183.1 | - | 4 - 5 | 957 | 62 | |
AK029531.1 | - | 4 - 5 | 417 | 62 | |
AK030423.1 | - | 4 - 5 | 975 | 62 | |
AK050499.1 | - | 5 - 6 | 848 | 62 | |
AK077123.1 | - | 3 - 4 | 341 | 62 | |
AK082180.1 | - | 4 - 5 | 960 | 62 | |
AK082497.1 | - | 4 - 5 | 758 | 62 | |
AK133243.1 | - | 5 - 6 | 519 | 62 | |
AK139923.1 | - | 3 - 4 | 920 | 62 | |
BB614593.1 | - | 3 - 4 | 393 | 62 | |
BC032277.1 | - | 4 - 5 | 902 | 62 | |
L76944.1 | - | 4 - 5 | 603 | 62 | |
L76946.1 | - | 5 - 6 | 949 | 62 | |
L76947.1 | - | 4 - 5 | 436 | 62 |