Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde1c
Entrez Gene ID
18575
Gene Name
phosphodiesterase 1C
Gene Aliases
-
Chromosome Location
Chr.6: 56069798 - 56652516 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025568.2 NP_001020739.1 7 - 8 1299 102
NM_001159952.1 NP_001153424.1 6 - 7 1272 102
NM_001159953.1 NP_001153425.1 7 - 8 1299 102
NM_001159955.1 NP_001153427.1 8 - 9 861 102
NM_001159956.1 NP_001153428.1 6 - 7 718 102
NM_001159957.1 NP_001153429.1 7 - 8 745 102
NM_001159960.1 NP_001153432.1 8 - 9 1173 102
NM_011054.4 NP_035184.1 8 - 9 1276 102
GenBank mRNA AK014887.1 - 6 - 7 689 102
AK015375.1 - 8 - 9 830 102
AK029531.1 - 7 - 8 743 102
AK030423.1 - 7 - 8 1301 102
AK050499.1 - 8 - 9 1174 102
AK077123.1 - 6 - 7 667 102
AK082180.1 - 7 - 8 1286 102
AK082497.1 - 7 - 8 1084 102
AK133243.1 - 8 - 9 845 102
AK139923.1 - 6 - 7 1246 102
BC032277.1 - 7 - 8 1228 102
L76944.1 - 7 - 8 929 102
L76946.1 - 8 - 9 1275 102
L76947.1 - 7 - 8 762 102

View Full Product Details