Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aloxe3
Entrez Gene ID
23801
Gene Name
arachidonate lipoxygenase 3
Gene Aliases
e-LOX-3, eLOX-3
Chromosome Location
Chr.11: 69125674 - 69149115 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011786.2 NP_035916.2 10 - 11 1420 104
GenBank mRNA AK141003.1 - 9 - 10 4186 104
BC116258.1 - 10 - 11 1419 104
BC116259.1 - 10 - 11 1419 104
Y14695.1 - 10 - 11 1420 104

View Full Product Details