The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289653.1 | NP_001276582.1 | 15 - 16 | 1618 | 70 |
GenBank mRNA | AK035019.1 | - | 13 - 14 | 1471 | 70 |
AK039790.1 | - | - | 1682 | 70 | |
AK048913.1 | - | - | 1889 | 70 | |
BC051968.1 | - | 14 - 15 | 1586 | 70 | |
U59230.1 | - | 15 - 16 | 1619 | 70 |