Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nptx2
Entrez Gene ID
53324
Gene Name
neuronal pentraxin 2
Gene Aliases
Narp, np2
Chromosome Location
Chr.5: 144545887 - 144557478 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016789.3 NP_058069.1 1 - 2 603 77
GenBank mRNA AF049124.1 - 1 - 2 493 77

View Full Product Details