The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018868.2 | NP_061356.2 | 12 - 13 | 1447 | 75 |
GenBank mRNA | AF053232.1 | - | 12 - 13 | 1352 | 75 |
AK034577.1 | - | - | 100 | 75 | |
AK044216.1 | - | - | 1444 | 75 | |
AK133959.1 | - | 12 - 13 | 1445 | 75 | |
AK134286.1 | - | 12 - 13 | 1448 | 75 | |
BC076604.1 | - | 12 - 13 | 1448 | 75 | |
BC085135.1 | - | 12 - 13 | 1417 | 75 |