Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Net1
Entrez Gene ID
56349
Gene Name
neuroepithelial cell transforming gene 1
Gene Aliases
Net1a, mNET1
Chromosome Location
Chr.13: 3882018 - 3918220 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019671.3 NP_062645.2 3 - 4 412 116
GenBank mRNA AF094520.1 - 3 - 4 308 116
CF167482.1 - 3 - 4 396 116

View Full Product Details